Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106196.1 | NP_001099666.1 | 9 | 810 | ||
XM_008772605.1 | XP_008770827.1 | 10 | 4674 | |||
GenBank mRNA | BC168926.1 | - | 928 |