Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136162.1 | NP_001129634.1 | 5 | 530 | ||
XM_006255802.2 | XP_006255864.1 | 7 | 698 | |||
XM_006255803.2 | XP_006255865.1 | 7 | 698 | |||
XM_006255805.2 | XP_006255867.1 | 5 | 539 |