Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231655.2 | XP_006231717.1 | 15 | 2228 | ||
XM_008760580.1 | XP_008758802.1 | 16 | 2334 | |||
XM_008760581.1 | XP_008758803.1 | 15 | 2249 | |||
GenBank mRNA | AB568258.1 | - | 13 | 1697 |