Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003357.4 | NP_003348.1 | 3 | 416 | ||
GenBank mRNA | BC004481.2 | - | 3 | 400 | 3688615 | |
DB196705.1 | - | 3 | 417 | |||
U01101.1 | - | 3 | 364 |