Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748874.3 | XP_003748922.1 | 3 | 293 |
Set Membership: |
G-protein coupled receptor |