Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297615.1 | NP_001284544.1 | 4 | 1086 | ||
NM_001297616.1 | NP_001284545.1 | 5 | 916 | |||
NM_021139.2 | NP_066962.2 | 4 | 1086 | |||
GenBank mRNA | AF064200.1 | - | 4 | 1064 | ||
AF081793.1 | - | 4 | 1074 | |||
AJ005162.1 | - | 4 | 1072 | |||
AK292748.1 | - | 4 | 1086 | |||
AK300084.1 | - | 5 | 916 | |||
AY529122.1 | - | 4 | 1039 | |||
BC026264.1 | - | 4 | 1083 | 4709436 | ||
Y00317.1 | - | 4 | 1076 |