Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC292543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025639.1 | NP_001020810.1 | 10 | 1652 | ||
XM_006228022.2 | XP_006228084.1 | 10 | 1744 | |||
XM_008758754.1 | XP_008756976.1 | 10 | 1817 | |||
GenBank mRNA | BC098062.1 | - | 10 | 1652 | 7458331 |