Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191620.1 | NP_001178549.1 | 4 | 1404 | ||
XM_006228177.2 | XP_006228239.1 | 3 | 1548 | |||
GenBank mRNA | FQ212491.1 | - | 4 | 1498 |
Gene Symbol: LOC103691059 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759084.1 | XP_008757306.1 | 1 | 624 |
Gene Symbol: LOC691135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759083.1 | XP_008757305.1 | 4 | 2269 |