Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106225.1 | NP_001099695.1 | 7, 8 | 969 | ||
XM_006228216.2 | XP_006228278.1 | 7, 8 | 993 | |||
GenBank mRNA | BC168158.1 | - | 7, 8 | 964 | 8364284 | |
FQ212233.1 | - | 4, 5 | 434 |