Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025640.1 | NP_001020811.1 | 2, 3 | 226 | ||
XM_006227646.2 | XP_006227708.1 | 3, 4 | 376 | |||
GenBank mRNA | BC098787.1 | - | 2, 3 | 226 | 7375248 |