Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sympk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100830.1 | NP_001094300.1 | 6 | 425 | ||
XM_008758908.1 | XP_008757130.1 | 6 | 675 |