Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008389.2 | NP_001008390.1 | 8 | 1939 | ||
NM_001278614.1 | NP_001265543.1 | 9 | 2101 | |||
NM_003361.3 | NP_003352.2 | 8 | 2002 | |||
XM_011545934.1 | XP_011544236.1 | 9 | 2080 | |||
XM_011545935.1 | XP_011544237.1 | 10 | 2139 | |||
XM_011545936.1 | XP_011544238.1 | 10 | 2076 | |||
XM_011545937.1 | XP_011544239.1 | 10 | 1966 | |||
XM_011545938.1 | XP_011544240.1 | 9 | 1998 | |||
XM_011545940.1 | XP_011544242.1 | 9 | 2016 | |||
XM_017023641.1 | XP_016879130.1 | 8 | 1939 | |||
GenBank mRNA | AK055722.1 | - | 9 | 1705 | ||
AK091961.1 | - | 9 | 1933 | |||
AK096043.1 | - | 9 | 1582 | |||
AK127643.1 | - | 9 | 1435 | |||
AK127648.1 | - | 9 | 1698 | |||
BC035975.1 | - | 9 | 1688 | 4556427 | ||
M15881.1 | - | 8 | 1878 | |||
M17778.1 | - | 7 | 1815 |