Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008303.1 | NP_001008304.1 | 6, 7 | 764 | ||
XM_006228564.2 | XP_006228626.1 | 6, 7 | 895 | |||
GenBank mRNA | BC086331.1 | - | 6, 7 | 764 | 7300752 | |
FQ227089.1 | - | 5, 6 | 658 | |||
FQ231860.1 | - | 4, 5 | 468 | |||
FQ233559.1 | - | 5, 6 | 658 |