Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008303.1 | NP_001008304.1 | 3, 4 | 332 | ||
XM_006228564.2 | XP_006228626.1 | 3, 4 | 463 | |||
GenBank mRNA | BC086331.1 | - | 3, 4 | 332 | 7300752 | |
FQ227089.1 | - | 2, 3 | 226 | |||
FQ231860.1 | - | 1, 2 | 36 | |||
FQ233559.1 | - | 2, 3 | 226 |