Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008389.2 | NP_001008390.1 | 7 | 1729 | ||
NM_001278614.1 | NP_001265543.1 | 8 | 1891 | |||
NM_003361.3 | NP_003352.2 | 7 | 1792 | |||
XM_011545934.1 | XP_011544236.1 | 8 | 1870 | |||
XM_011545935.1 | XP_011544237.1 | 9 | 1929 | |||
XM_011545936.1 | XP_011544238.1 | 9 | 1866 | |||
XM_011545937.1 | XP_011544239.1 | 9 | 1756 | |||
XM_011545938.1 | XP_011544240.1 | 8 | 1788 | |||
XM_011545940.1 | XP_011544242.1 | 8 | 1806 | |||
XM_017023641.1 | XP_016879130.1 | 7 | 1729 | |||
GenBank mRNA | AK091961.1 | - | 8 | 1723 | ||
AK096043.1 | - | 8 | 1372 | |||
AK127643.1 | - | 8 | 1225 | |||
AK127648.1 | - | 8 | 1488 | |||
BC035975.1 | - | 8 | 1478 | 4556427 | ||
M15881.1 | - | 7 | 1668 | |||
M17778.1 | - | 6 | 1605 |