Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blvrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106236.1 | NP_001099706.1 | 2 | 240 | ||
XM_008759117.1 | XP_008757339.1 | 2 | 161 | |||
GenBank mRNA | BC168943.1 | - | 2 | 144 | 7322093 | |
FQ228106.1 | - | 2 | 124 | |||
FQ230737.1 | - | 2 | 187 | |||
FQ232598.1 | - | 2 | 187 |