Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012474.4 | NP_036606.2 | 2 | 534 | ||
XM_017002236.1 | XP_016857725.1 | 3 | 739 | |||
GenBank mRNA | AB062451.1 | - | 2 | 153 | ||
AF236637.1 | - | 2 | 213 | |||
AK294537.1 | - | 2 | 207 | |||
BC002906.2 | - | 2 | 374 | 3940564 | ||
DR762696.1 | - | 2 | 534 |