Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106240.1 | NP_001099710.1 | 9 | 839 | ||
GenBank mRNA | FQ212522.1 | - | 9 | 841 |