Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000373.3 | NP_000364.1 | 3 | 922 | ||
NR_033434.1 | 2 | 768 | ||||
NR_033437.1 | 4 | 1021 | ||||
GenBank mRNA | AB062285.1 | - | 3 | 844 | ||
AK291306.1 | - | 3 | 838 | |||
AL080099.2 | - | 2 | 701 | |||
BC000364.2 | - | 3 | 839 | 2964556 | ||
BC007511.2 | - | 3 | 839 | 2964556 | ||
CR456787.1 | - | 3 | 816 | |||
D86227.1 | - | 3 | 835 | |||
D86228.1 | - | 3 | 835 | |||
D86230.1 | - | 3 | 835 | |||
EU921886.1 | - | 3 | 836 | |||
EU921887.1 | - | 2 | 682 | |||
EU921888.1 | - | 2 | 682 | |||
EU921889.1 | - | 3 | 781 | |||
EU921890.1 | - | 3 | 836 | |||
EU921891.1 | - | 4 | 935 | |||
EU921893.1 | - | 3 | 489 | |||
EU921894.1 | - | 3 | 836 | |||
EU921895.1 | - | 2 | 682 | |||
J03626.1 | - | 3 | 920 | |||
KU178375.1 | - | 3 | 816 | |||
KU178376.1 | - | 4 | 915 | |||
M36661.1 | - | 3 | 782 |