Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102550585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228694.2 | XP_006228756.1 | 6 | 1294 | ||
XM_006228695.2 | XP_006228757.1 | 1278 | ||||
XM_008759220.1 | XP_008757442.1 | 1313 |
Gene Symbol: Catsperg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170340.1 | NP_001163811.1 | 6 | 687 | ||
XM_006228722.2 | XP_006228784.1 | 6 | 1273 | |||
XM_008759124.1 | XP_008757346.1 | 6 | 1306 | |||
XM_008759125.1 | XP_008757347.1 | 6 | 1288 | |||
XM_008759126.1 | XP_008757348.1 | 6 | 1306 | |||
XM_008759127.1 | XP_008757349.1 | 5 | 504 |