Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000373.3 | NP_000364.1 | 2, 3 | 413 | ||
NR_033437.1 | 3, 4 | 512 | ||||
GenBank mRNA | AB062285.1 | - | 2, 3 | 335 | ||
AK291306.1 | - | 2, 3 | 329 | |||
BC000364.2 | - | 2, 3 | 330 | 2964556 | ||
BC007511.2 | - | 2, 3 | 330 | 2964556 | ||
CR456787.1 | - | 2, 3 | 307 | |||
D86227.1 | - | 2, 3 | 326 | |||
D86228.1 | - | 2, 3 | 326 | |||
D86230.1 | - | 2, 3 | 326 | |||
EU921886.1 | - | 2, 3 | 327 | |||
EU921890.1 | - | 2, 3 | 327 | |||
EU921891.1 | - | 3, 4 | 426 | |||
EU921892.1 | - | 2, 3 | 327 | |||
EU921894.1 | - | 2, 3 | 327 | |||
J03626.1 | - | 2, 3 | 411 | |||
KU178375.1 | - | 2, 3 | 307 | |||
KU178376.1 | - | 3, 4 | 406 | |||
M36661.1 | - | 2, 3 | 273 |