Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL14A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021110.3 | NP_066933.1 | 14 | 1961 | ||
XM_005251059.3 | XP_005251116.1 | 14 | 1961 | |||
XM_006716651.3 | XP_006716714.1 | 14 | 1824 | |||
XM_017013809.1 | XP_016869298.1 | 14 | 1750 | |||
GenBank mRNA | BC140893.1 | - | 13 | 1730 | 9021761 | |
BX472674.1 | - | 3 | 280 | |||
M64108.1 | - | 9 | 1130 | |||
M64109.1 | - | 962 |