Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170405.1 | NP_001163876.1 | 4 | 441 | ||
XM_008759331.1 | XP_008757553.1 | 3 | 360 |