Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748848.3 | XP_003748896.1 | 744 |
Gene Symbol: Klk1c10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135173.1 | NP_001128645.1 | 724 | |||
GenBank mRNA | S48142.1 | - | 4 | 681 |