Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106259.3 | NP_001099729.2 | 4 | 697 | ||
XM_006229003.2 | XP_006229065.1 | 4 | 1173 | |||
XM_006229004.2 | XP_006229066.1 | 4 | 873 | |||
XM_006229005.2 | XP_006229067.1 | 4 | 849 | |||
XM_008759335.1 | XP_008757557.1 | 3 | 426 | |||
GenBank mRNA | EV773331.1 | - | 3 | 613 |