Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047853.1 | NP_001041318.1 | 4 | 345 | ||
XM_006229367.2 | XP_006229429.1 | 5 | 887 | |||
XM_006229368.2 | XP_006229430.1 | 5 | 887 | |||
GenBank mRNA | AY325151.1 | - | 4 | 345 | ||
AY539868.1 | - | 3 | 305 | |||
FQ222682.1 | - | 5 | 767 |