Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008510.1 | NP_001008510.1 | 3 | 518 | ||
GenBank mRNA | BC086557.1 | - | 3 | 518 | 7300717 | |
FQ233960.1 | - | 4 | 575 | |||
FQ234031.1 | - | 585 | ||||
FQ234316.1 | - | 3 | 571 |