Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199975.2 | NP_001186904.1 | 3 | 265 | ||
NM_001254752.1 | NP_001241681.1 | 2 | 151 | |||
NM_006294.4 | NP_006285.1 | 2 | 151 | |||
NR_045639.1 | 2 | 151 | ||||
GenBank mRNA | AK295007.1 | - | 2 | 63 | ||
AK311756.1 | - | 2 | 76 | |||
BC005230.1 | - | 2 | 100 | 3961169 | ||
BI547559.1 | - | 2 | 152 | |||
BU535281.1 | - | 2 | 59 | |||
CR542176.1 | - | 1 | 47 | |||
CR542196.1 | - | 2 | 47 | |||
M22348.1 | - | 2 | 79 | |||
M26700.1 | - | 2 | 99 | |||
X13585.1 | - | 2 | 79 |
Gene Symbol: UQCRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002308.1 | 1 | 54 |