Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam154b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106277.1 | NP_001099747.1 | 3 | 1014 | ||
XM_006229473.2 | XP_006229535.1 | 4 | 1225 | |||
XM_006229474.2 | XP_006229536.1 | 4 | 2917 | |||
XM_006229475.2 | XP_006229537.1 | 5 | 1308 | |||
XM_008759511.1 | XP_008757733.1 | 5 | 2116 |