Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170432.1 | NP_001163903.1 | 22 | 2973 | ||
XM_008759610.1 | XP_008757832.1 | 23 | 3200 |