Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106283.1 | NP_001099753.1 | 5 | 650 | ||
XM_006229858.2 | XP_006229920.1 | 4 | 530 | |||
XM_006229859.2 | XP_006229921.1 | 5 | 732 | |||
XM_008759703.1 | XP_008757925.1 | 4 | 565 | |||
GenBank mRNA | FQ211323.1 | - | 4 | 497 |