Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000131.1 | NP_001000131.1 | 1 | 649 |
Gene Symbol: Olr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000134.1 | NP_001000134.1 | 1 | 649 |