Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000133.1 | NP_001000133.1 | 1 | 663 |
Gene Symbol: Olr67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001279.1 | NP_001001279.1 | 1 | 633 |