Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001271.1 | NP_001001271.1 | 1 | 636 |
Gene Symbol: Olr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000136.1 | NP_001000136.1 | 1 | 636 |