Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276373.1 | NP_001263302.1 | 6 | 529 | ||
NM_007122.4 | NP_009053.1 | 6 | 625 | |||
NM_207005.2 | NP_996888.1 | 6 | 594 | |||
GenBank mRNA | AB098540.1 | - | 4 | 387 | ||
AK314876.1 | - | 6 | 545 | |||
AL832119.1 | - | 6 | 662 | |||
BC035505.2 | - | 6 | 573 | 4559819 | ||
KU178379.1 | - | 4 | 418 | |||
KU178380.1 | - | 4 | 387 | |||
X55666.1 | - | 6 | 538 |