Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000154.1 | NP_001000154.1 | 1 | 570 |
Gene Symbol: Olr114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000951.1 | NP_001000951.1 | 1 | 570 |