Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqlnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013899.1 | NP_001013921.1 | 1 | 1917 | ||
GenBank mRNA | BC083634.1 | - | 1 | 1917 | 7134647 |