Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000173.1 | NP_001000173.1 | 1 | 302 |
Gene Symbol: Olr162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000950.1 | NP_001000950.1 | 1 | 302 |