Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000180.1 | NP_001000180.1 | 1 | 360 |
Gene Symbol: Olr184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000182.1 | NP_001000182.1 | 1 | 360 |