Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UTY SIRNA Assays ›
Gene Symbol:
UTY
Gene Name:
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Gene Aliases:
KDM6AL, UTY1
Chromosome Location:
Chr.Y: 13230770 - 13480670 on Build GRCh38
UniGene ID:
Hs.115277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14739
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UTY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258249.1
NP_001245178.1
17
2759
NM_001258251.1
NP_001245180.1
15
2468
NM_001258252.1
NP_001245181.1
16
2603
NM_001258253.1
NP_001245182.1
15
2468
NM_001258256.1
NP_001245185.1
16
2603
NM_001258258.1
NP_001245187.1
16
2624
NM_001258259.1
NP_001245188.1
15
2420
NM_001258260.1
NP_001245189.1
16
2558
NM_001258261.1
NP_001245190.1
16
2594
NM_001258263.1
NP_001245192.1
15
2468
NM_001258264.1
NP_001245193.1
16
2555
NM_001258265.1
NP_001245194.1
11
2219
NM_001258266.1
NP_001245195.1
16
2603
NM_001258267.1
NP_001245196.1
15
2468
NM_007125.4
NP_009056.3
15
2468
NM_182659.1
NP_872600.1
15
2468
NM_182660.1
NP_872601.1
15
2468
NR_047596.1
15
2468
NR_047597.1
16
2574
NR_047598.1
17
2704
NR_047599.1
15
2468
NR_047601.1
16
2603
NR_047602.1
14
2433
NR_047603.1
16
2591
NR_047605.1
15
2420
NR_047606.1
16
2604
NR_047607.1
17
2814
NR_047608.1
16
2555
NR_047610.1
15
2468
NR_047611.1
16
2603
NR_047612.1
17
2759
NR_047613.1
16
2709
NR_047614.1
17
2672
NR_047615.1
16
2526
NR_047616.1
16
2582
NR_047617.1
17
2730
NR_047619.1
16
2526
NR_047620.1
16
2603
NR_047621.1
17
2830
NR_047622.1
16
2660
NR_047623.1
16
2603
NR_047624.1
17
2688
NR_047625.1
14
2341
NR_047626.1
15
2468
NR_047627.1
14
2433
NR_047628.1
15
2468
NR_047629.1
15
2468
NR_047630.1
15
2468
NR_047631.1
15
2468
NR_047634.1
15
2589
NR_047635.1
15
2463
NR_047636.1
16
2558
NR_047637.1
16
2526
NR_047638.1
15
2534
NR_047639.1
14
2374
NR_047640.1
16
2574
NR_047641.1
15
2468
NR_047642.1
15
2468
NR_047643.1
16
2603
NR_047644.1
17
2661
NR_047645.1
16
2674
NR_047646.1
14
2341
NR_047647.1
15
2425
XM_005262518.3
XP_005262575.1
16
1949
XM_006724875.3
XP_006724938.1
17
2105
XM_011531441.2
XP_011529743.1
17
2105
XM_011531442.2
XP_011529744.1
17
2105
XM_011531443.2
XP_011529745.1
17
2039
XM_011531445.2
XP_011529747.1
16
1970
XM_011531446.2
XP_011529748.1
16
1949
XM_011531447.2
XP_011529749.1
16
1970
XM_011531448.2
XP_011529750.1
16
1904
XM_011531449.2
XP_011529751.1
16
1949
XM_011531450.2
XP_011529752.1
16
1904
XM_011531451.2
XP_011529753.1
15
1814
XM_011531452.2
XP_011529754.1
15
1814
XM_011531453.2
XP_011529755.1
17
2105
XM_011531454.2
XP_011529756.1
17
2105
XM_011531455.2
XP_011529757.1
17
2105
XM_011531457.2
XP_011529759.1
17
2105
XM_011531458.2
XP_011529760.1
17
2039
XM_011531459.2
XP_011529761.1
17
2105
XM_011531460.2
XP_011529762.1
17
2105
XM_011531461.2
XP_011529763.1
17
2105
XM_011531462.2
XP_011529764.1
17
2039
XM_011531463.2
XP_011529765.1
16
1949
XM_011531464.2
XP_011529766.1
16
1949
XM_011531465.2
XP_011529767.1
15
1814
XM_017030066.1
XP_016885555.1
17
2039
XM_017030067.1
XP_016885556.1
16
1949
XM_017030068.1
XP_016885557.1
15
1814
XM_017030069.1
XP_016885558.1
15
1814
XM_017030070.1
XP_016885559.1
17
2105
XM_017030071.1
XP_016885560.1
17
2105
XM_017030072.1
XP_016885561.1
16
1904
XM_017030073.1
XP_016885562.1
15
1814
XM_017030074.1
XP_016885563.1
17
2039
GenBank mRNA
AF000994.1
-
15
2468
AF000995.1
-
15
2468
AF000996.1
-
15
2468
AK292858.1
-
15
1718
AK308952.1
-
1
67
AY682463.1
-
14
1370
AY730613.1
-
16
1619
AY769061.1
-
17
1699
AY769062.1
-
16
1569
AY769063.1
-
15
1494
AY769064.1
-
14
1428
AY769065.1
-
15
1463
AY769066.1
-
17
1754
AY769067.1
-
15
1463
AY769068.1
-
16
1521
AY769069.1
-
14
1428
AY934848.1
-
15
1463
AY934849.1
-
16
1569
AY934851.1
-
15
1415
AY934852.1
-
15
1454
AY934853.1
-
15
1444
AY934854.1
-
14
1364
AY934855.1
-
14
1364
AY934856.1
-
14
1380
AY934857.1
-
16
1577
CR936684.1
-
7
566
DQ140388.1
-
15
1463
DQ140389.1
-
16
1598
DQ341385.1
-
15
1463
DQ341386.1
-
15
1463
DQ341387.1
-
14
1364
DQ345542.1
-
16
1653
DQ345543.1
-
16
1553
DQ345544.1
-
17
1688
DQ350457.1
-
15
1463
DQ350458.1
-
16
1577
DQ350459.1
-
15
1463
EF612661.1
-
14
1364
EF612663.1
-
16
1598
EF612665.1
-
15
1415
EF612667.1
-
14
1364
EF612668.1
-
17
1754
EF612670.1
-
13
1271
EF612672.1
-
15
1415
EF612673.1
-
16
1619
EF612674.1
-
15
1415
EF612675.1
-
16
1550
EF612677.1
-
17
1754
EF612680.1
-
16
1553
EF612683.1
-
16
1589
EF612685.1
-
17
1688
EF612686.1
-
16
1598
EF612687.1
-
15
1511
EF612689.1
-
16
1598
EF612690.1
-
14
1364
EF612691.1
-
16
1598
EF612692.1
-
13
1373
EF612693.1
-
15
1463
EF612695.1
-
16
1550
EF612696.1
-
16
1553
EF612697.1
-
15
1463
EF612698.1
-
13
1373
EF653165.1
-
14
1428
EF653166.1
-
14
1369
EF653167.1
-
13
1329
EF653168.1
-
16
1598
EF653169.1
-
14
1428
EF653170.1
-
13
1329
EF653171.1
-
15
1463
EF653172.1
-
14
1354
EF653173.1
-
15
1463
EF653174.1
-
17
1659
EF653175.1
-
17
1704
EF653176.1
-
14
1413
EF653178.1
-
18
1820
EF653179.1
-
15
1463
EF653180.1
-
16
1569
EF653181.1
-
16
1553
EF653182.1
-
14
1428
EF653183.1
-
16
1569
EF653184.1
-
16
1577
EF653186.1
-
16
1586
EF653189.1
-
11
1214
EF653190.1
-
16
1598
EF653192.1
-
14
1369
EF653193.1
-
16
1569
EF653194.1
-
16
1543
EF653195.1
-
16
1569
EF653196.1
-
15
1463
EF653197.1
-
14
1428
EF653198.1
-
15
1463
EF653199.1
-
17
1683
EF653200.1
-
16
1598
EF653201.1
-
15
1504
EF653202.1
-
13
1242
EF653204.1
-
16
1579
EF653205.1
-
15
1463
EF653207.1
-
17
1656
EF653208.1
-
16
1569
EF653209.1
-
16
1669
EF653211.1
-
17
1627
EF653212.1
-
15
1463
EF653213.1
-
14
1336
EF653214.1
-
17
1780
EF653215.1
-
15
1533
EF653216.1
-
15
1515
EF653217.1
-
15
1420
EF653218.1
-
15
1415
EF653219.1
-
17
1754
EF653221.1
-
13
1329
EF653223.1
-
16
1543
EF653224.1
-
17
1809
EF685285.1
-
16
1550
EF685286.1
-
16
1598
EF685287.1
-
15
1463
EF685288.1
-
17
1754
EF685289.1
-
8
850
EF685290.1
-
16
1704
EF685291.1
-
17
1667
EF685292.1
-
16
1521
EF685293.1
-
16
1577
EF685294.1
-
17
1725
EF685296.1
-
16
1521
EF685297.1
-
16
1598
EF685298.1
-
17
1825
EF685299.1
-
16
1569
EF685300.1
-
16
1655
EF685301.1
-
16
1598
EF685302.1
-
15
1436
EF685303.1
-
15
1463
EF685304.1
-
15
1486
EF685305.1
-
16
1538
EF685306.1
-
15
1499
EF685307.1
-
17
1683
EF685308.1
-
14
1336
EF685309.1
-
15
1463
EF685310.1
-
15
1463
EF685311.1
-
15
1463
EF685312.1
-
14
1428
EF685314.1
-
15
1463
EF685315.1
-
14
1369
EF685316.1
-
15
1463
EF685317.1
-
16
1619
EF685318.1
-
14
1364
EF685319.1
-
14
1364
EF685320.1
-
15
1415
EF685321.1
-
15
1463
EF685322.1
-
14
1364
EF685323.1
-
14
1364
EF685324.1
-
14
1280
EF685325.1
-
15
1463
EF685326.1
-
16
1521
EF685327.1
-
16
1538
EF685328.1
-
14
1364
EF685331.1
-
1463
EF685332.1
-
16
1598
EF685333.1
-
17
1589
EF685334.1
-
15
1463
EF685335.1
-
11
1147
EF685338.1
-
15
1293
EF685339.1
-
14
1364
EF685340.1
-
14
1364
EF685341.1
-
14
1364
EF685342.1
-
15
1463
EU026275.1
-
15
1470
EU026277.1
-
16
1529
EU026279.1
-
13
1325
EU026280.1
-
17
1679
EU026281.1
-
14
1428
EU026282.1
-
13
1270
EU026284.1
-
15
1459
EU026286.1
-
14
1380
EU026287.1
-
13
1242
EU026289.1
-
14
1370
EU026290.1
-
15
1664
EU026295.1
-
13
1325
EU026297.1
-
16
1565
EU026298.1
-
15
1494
EU026299.1
-
14
1428
EU026301.1
-
16
1600
EU026302.1
-
17
1727
EU026303.1
-
16
1593
EU026305.1
-
14
1364
EU026306.1
-
16
1577
EU026307.1
-
15
1463
EU026308.1
-
14
1435
EU026309.1
-
17
1640
EU026310.1
-
15
1463
EU026311.1
-
16
1598
EU026313.1
-
15
1415
EU026314.1
-
17
1704
EU026315.1
-
15
1494
EU026318.1
-
13
1330
EU026319.1
-
16
1521
EU026320.1
-
18
1762
EU026321.1
-
16
1593
EU026322.1
-
15
1458
EU026323.1
-
15
1558
EU026324.1
-
13
1253
EU026326.1
-
16
1569
EU026327.1
-
15
1534
EU026328.1
-
15
1518
EU026329.1
-
16
1624
EU026330.1
-
13
1368
EU026331.1
-
15
1463
EU026332.1
-
13
1329
EU026333.1
-
16
1627
EU026334.1
-
14
1361
EU026336.1
-
10
1094
EU026337.1
-
12
1202
EU026338.1
-
15
1463
EU026339.1
-
13
1329
EU026340.1
-
15
1549
EU026341.1
-
8
862
EU026342.1
-
15
1571
EU026343.1
-
16
1585
EU026345.1
-
15
1458
EU026346.1
-
13
1314
EU026347.1
-
13
1329
EU026348.1
-
15
1463
EU026349.1
-
15
1463
EU026350.1
-
14
1359
EU026351.1
-
15
1463
EU116343.1
-
10
991
EU116345.1
-
14
1364
EU116346.1
-
15
1463
EU116347.1
-
14
1469
EU116348.1
-
15
1464
EU116349.1
-
15
1557
EU116351.1
-
9
927
EU116352.1
-
15
1458
EU116353.1
-
16
1553
EU163941.1
-
15
1529
EU163942.1
-
16
1521
EU163943.1
-
6
686
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of gene expression
oxidation-reduction process
histone H3-K27 demethylation
histone demethylase activity
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K27 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-gx66z:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline