Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UTY SIRNA Assays ›
Gene Symbol:
UTY
Gene Name:
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Gene Aliases:
KDM6AL, UTY1
Chromosome Location:
Chr.Y: 13230770 - 13480670 on Build GRCh38
UniGene ID:
Hs.115277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14740
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UTY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258249.1
NP_001245178.1
5
1418
NM_001258250.1
NP_001245179.1
5
1418
NM_001258251.1
NP_001245180.1
5
1418
NM_001258252.1
NP_001245181.1
5
1418
NM_001258253.1
NP_001245182.1
5
1418
NM_001258254.1
NP_001245183.1
5
1418
NM_001258255.1
NP_001245184.1
5
1418
NM_001258256.1
NP_001245185.1
5
1418
NM_001258257.1
NP_001245186.1
5
1418
NM_001258258.1
NP_001245187.1
5
1418
NM_001258259.1
NP_001245188.1
5
1418
NM_001258260.1
NP_001245189.1
5
1418
NM_001258261.1
NP_001245190.1
5
1418
NM_001258262.1
NP_001245191.1
5
1418
NM_001258263.1
NP_001245192.1
5
1418
NM_001258264.1
NP_001245193.1
5
1418
NM_001258265.1
NP_001245194.1
3
1259
NM_001258266.1
NP_001245195.1
5
1418
NM_001258267.1
NP_001245196.1
5
1418
NM_001258268.1
NP_001245197.1
5
1418
NM_001258269.1
NP_001245198.1
5
1418
NM_001258270.1
NP_001245199.1
5
1418
NM_007125.4
NP_009056.3
5
1418
NM_182659.1
NP_872600.1
5
1418
NM_182660.1
NP_872601.1
5
1418
NR_047596.1
5
1418
NR_047597.1
5
1418
NR_047598.1
5
1418
NR_047599.1
5
1418
NR_047600.1
5
1418
NR_047601.1
5
1418
NR_047602.1
5
1418
NR_047603.1
6
1541
NR_047604.1
5
1418
NR_047605.1
5
1418
NR_047606.1
5
1418
NR_047607.1
5
1418
NR_047608.1
5
1418
NR_047609.1
5
1418
NR_047610.1
5
1418
NR_047611.1
5
1418
NR_047612.1
5
1418
NR_047613.1
4
1368
NR_047614.1
5
1418
NR_047615.1
5
1418
NR_047616.1
5
1418
NR_047617.1
5
1418
NR_047618.1
5
1418
NR_047619.1
5
1418
NR_047620.1
5
1418
NR_047621.1
5
1418
NR_047622.1
5
1418
NR_047623.1
5
1418
NR_047624.1
5
1418
NR_047625.1
5
1418
NR_047626.1
5
1418
NR_047627.1
5
1418
NR_047628.1
5
1418
NR_047629.1
5
1418
NR_047630.1
5
1418
NR_047631.1
5
1418
NR_047632.1
6
1498
NR_047633.1
5
1418
NR_047634.1
5
1418
NR_047635.1
5
1418
NR_047636.1
5
1418
NR_047637.1
5
1418
NR_047638.1
5
1418
NR_047639.1
5
1418
NR_047640.1
5
1418
NR_047641.1
5
1418
NR_047642.1
5
1418
NR_047643.1
5
1418
NR_047644.1
5
1418
NR_047645.1
5
1418
NR_047646.1
5
1418
NR_047647.1
5
1418
XM_005262518.3
XP_005262575.1
5
764
XM_006724875.3
XP_006724938.1
5
764
XM_011531441.2
XP_011529743.1
5
764
XM_011531442.2
XP_011529744.1
5
764
XM_011531443.2
XP_011529745.1
5
764
XM_011531445.2
XP_011529747.1
5
764
XM_011531446.2
XP_011529748.1
5
764
XM_011531447.2
XP_011529749.1
5
764
XM_011531448.2
XP_011529750.1
5
764
XM_011531449.2
XP_011529751.1
5
764
XM_011531450.2
XP_011529752.1
5
764
XM_011531451.2
XP_011529753.1
5
764
XM_011531452.2
XP_011529754.1
5
764
XM_011531453.2
XP_011529755.1
5
764
XM_011531454.2
XP_011529756.1
5
764
XM_011531455.2
XP_011529757.1
5
764
XM_011531457.2
XP_011529759.1
5
764
XM_011531458.2
XP_011529760.1
5
764
XM_011531459.2
XP_011529761.1
5
764
XM_011531460.2
XP_011529762.1
5
764
XM_011531461.2
XP_011529763.1
5
764
XM_011531462.2
XP_011529764.1
5
764
XM_011531463.2
XP_011529765.1
5
764
XM_011531464.2
XP_011529766.1
5
764
XM_011531465.2
XP_011529767.1
5
764
XM_017030066.1
XP_016885555.1
5
764
XM_017030067.1
XP_016885556.1
5
764
XM_017030068.1
XP_016885557.1
5
764
XM_017030069.1
XP_016885558.1
5
764
XM_017030070.1
XP_016885559.1
5
764
XM_017030071.1
XP_016885560.1
5
764
XM_017030072.1
XP_016885561.1
5
764
XM_017030073.1
XP_016885562.1
5
764
XM_017030074.1
XP_016885563.1
5
764
GenBank mRNA
AF000994.1
-
5
1418
AF000995.1
-
5
1418
AF000996.1
-
5
1418
AK292858.1
-
5
668
AY682463.1
-
5
419
AY730613.1
-
5
413
AY769061.1
-
5
413
AY769062.1
-
5
413
AY769064.1
-
5
413
AY769065.1
-
5
413
AY769066.1
-
5
413
AY769067.1
-
5
413
AY769068.1
-
5
413
AY769069.1
-
5
413
AY934848.1
-
5
413
AY934849.1
-
5
413
AY934850.1
-
5
413
AY934851.1
-
5
413
AY934852.1
-
5
413
AY934853.1
-
6
493
AY934854.1
-
5
413
AY934855.1
-
5
413
AY934856.1
-
5
413
AY934857.1
-
5
413
DQ140388.1
-
5
413
DQ140389.1
-
5
413
DQ140390.1
-
5
413
DQ341385.1
-
5
413
DQ341386.1
-
5
413
DQ341387.1
-
5
413
DQ345542.1
-
5
413
DQ345543.1
-
5
413
DQ345544.1
-
5
413
DQ350456.1
-
5
413
DQ350457.1
-
5
413
DQ350458.1
-
5
413
DQ350459.1
-
5
413
EF612660.1
-
5
413
EF612661.1
-
5
413
EF612662.1
-
5
413
EF612663.1
-
5
413
EF612664.1
-
5
413
EF612665.1
-
5
413
EF612666.1
-
5
413
EF612667.1
-
5
413
EF612668.1
-
5
413
EF612669.1
-
5
413
EF612670.1
-
5
413
EF612671.1
-
5
413
EF612672.1
-
5
413
EF612673.1
-
5
413
EF612674.1
-
5
413
EF612675.1
-
5
413
EF612676.1
-
5
413
EF612677.1
-
5
413
EF612678.1
-
5
413
EF612679.1
-
5
413
EF612680.1
-
5
413
EF612681.1
-
5
413
EF612682.1
-
5
413
EF612683.1
-
5
413
EF612684.1
-
5
413
EF612685.1
-
5
413
EF612686.1
-
5
413
EF612687.1
-
5
413
EF612688.1
-
5
413
EF612689.1
-
5
413
EF612690.1
-
5
413
EF612691.1
-
5
413
EF612692.1
-
5
413
EF612693.1
-
5
413
EF612694.1
-
5
413
EF612695.1
-
5
413
EF612696.1
-
5
413
EF612697.1
-
5
413
EF612698.1
-
5
413
EF612699.1
-
5
413
EF653165.1
-
5
413
EF653166.1
-
5
413
EF653167.1
-
5
413
EF653168.1
-
5
413
EF653169.1
-
5
413
EF653170.1
-
5
413
EF653171.1
-
5
413
EF653172.1
-
4
304
EF653173.1
-
5
413
EF653174.1
-
5
413
EF653176.1
-
4
363
EF653177.1
-
5
413
EF653178.1
-
5
413
EF653179.1
-
5
413
EF653181.1
-
5
413
EF653182.1
-
5
413
EF653183.1
-
5
413
EF653184.1
-
5
413
EF653185.1
-
5
413
EF653186.1
-
6
536
EF653187.1
-
5
413
EF653189.1
-
3
254
EF653190.1
-
5
413
EF653191.1
-
5
413
EF653192.1
-
5
413
EF653193.1
-
5
413
EF653195.1
-
5
413
EF653196.1
-
5
413
EF653197.1
-
5
413
EF653198.1
-
5
413
EF653199.1
-
5
413
EF653200.1
-
5
413
EF653201.1
-
5
413
EF653202.1
-
5
413
EF653203.1
-
5
413
EF653204.1
-
5
413
EF653205.1
-
5
413
EF653206.1
-
5
413
EF653207.1
-
5
413
EF653208.1
-
5
413
EF653209.1
-
5
413
EF653211.1
-
5
413
EF653212.1
-
5
413
EF653213.1
-
5
413
EF653215.1
-
5
413
EF653216.1
-
5
413
EF653217.1
-
5
413
EF653218.1
-
5
413
EF653219.1
-
5
413
EF653220.1
-
5
413
EF653221.1
-
5
413
EF653222.1
-
6
493
EF653223.1
-
6
493
EF653224.1
-
5
413
EF685283.1
-
5
413
EF685284.1
-
5
413
EF685285.1
-
5
413
EF685286.1
-
5
413
EF685287.1
-
5
413
EF685290.1
-
4
363
EF685291.1
-
5
413
EF685292.1
-
5
413
EF685293.1
-
5
413
EF685294.1
-
5
413
EF685295.1
-
5
413
EF685296.1
-
5
413
EF685297.1
-
5
413
EF685298.1
-
5
413
EF685299.1
-
5
413
EF685300.1
-
5
413
EF685301.1
-
5
413
EF685302.1
-
5
413
EF685303.1
-
5
413
EF685304.1
-
6
535
EF685305.1
-
6
488
EF685306.1
-
5
413
EF685307.1
-
5
413
EF685308.1
-
5
413
EF685309.1
-
5
413
EF685310.1
-
5
413
EF685311.1
-
5
413
EF685312.1
-
5
413
EF685313.1
-
5
413
EF685314.1
-
5
413
EF685315.1
-
5
413
EF685316.1
-
5
413
EF685317.1
-
5
413
EF685318.1
-
5
413
EF685319.1
-
5
413
EF685320.1
-
5
413
EF685321.1
-
5
413
EF685322.1
-
5
413
EF685323.1
-
5
413
EF685324.1
-
5
413
EF685325.1
-
5
413
EF685326.1
-
5
413
EF685327.1
-
6
488
EF685329.1
-
5
413
EF685330.1
-
5
413
EF685331.1
-
413
EF685332.1
-
5
413
EF685333.1
-
5
413
EF685334.1
-
5
413
EF685335.1
-
5
413
EF685336.1
-
5
413
EF685337.1
-
5
413
EF685338.1
-
5
413
EF685339.1
-
5
413
EF685340.1
-
5
413
EF685341.1
-
5
413
EF685342.1
-
5
413
EU026275.1
-
5
413
EU026276.1
-
5
413
EU026277.1
-
5
413
EU026278.1
-
5
413
EU026279.1
-
5
413
EU026280.1
-
5
413
EU026281.1
-
5
413
EU026284.1
-
5
413
EU026285.1
-
5
413
EU026286.1
-
5
413
EU026287.1
-
5
413
EU026288.1
-
5
413
EU026289.1
-
5
413
EU026290.1
-
5
413
EU026292.1
-
5
413
EU026294.1
-
5
413
EU026295.1
-
5
413
EU026296.1
-
5
413
EU026298.1
-
5
413
EU026299.1
-
5
413
EU026300.1
-
5
413
EU026301.1
-
5
413
EU026302.1
-
5
413
EU026303.1
-
5
413
EU026304.1
-
5
413
EU026305.1
-
5
413
EU026306.1
-
5
413
EU026307.1
-
5
413
EU026308.1
-
5
413
EU026309.1
-
5
413
EU026310.1
-
5
413
EU026311.1
-
5
413
EU026313.1
-
5
413
EU026314.1
-
5
413
EU026315.1
-
5
413
EU026316.1
-
6
493
EU026317.1
-
5
413
EU026318.1
-
4
363
EU026319.1
-
5
413
EU026320.1
-
5
413
EU026321.1
-
5
413
EU026322.1
-
5
413
EU026323.1
-
5
413
EU026324.1
-
5
413
EU026325.1
-
5
413
EU026326.1
-
5
413
EU026327.1
-
5
413
EU026328.1
-
5
413
EU026329.1
-
5
413
EU026330.1
-
5
413
EU026331.1
-
5
413
EU026332.1
-
5
413
EU026333.1
-
5
413
EU026334.1
-
5
413
EU026335.1
-
5
413
EU026338.1
-
5
413
EU026339.1
-
5
413
EU026340.1
-
5
413
EU026342.1
-
4
363
EU026343.1
-
6
535
EU026344.1
-
5
413
EU026345.1
-
5
413
EU026346.1
-
4
363
EU026347.1
-
5
413
EU026348.1
-
5
413
EU026349.1
-
5
413
EU026350.1
-
5
413
EU026351.1
-
5
413
EU116345.1
-
5
413
EU116346.1
-
5
413
EU116347.1
-
5
413
EU116348.1
-
5
293
EU116349.1
-
6
535
EU116350.1
-
4
283
EU116352.1
-
5
413
EU116353.1
-
5
413
EU163941.1
-
5
413
EU163942.1
-
5
413
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008773696, XM_008773695, FJ775728.
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of gene expression
oxidation-reduction process
histone H3-K27 demethylation
histone demethylase activity
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K27 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline