Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVRAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003369.3 | NP_003360.2 | 13, 14 | 1554 | ||
XM_011545226.2 | XP_011543528.1 | 13, 14 | 1869 | |||
XM_017018224.1 | XP_016873713.1 | 11, 12 | 1308 | |||
XM_017018225.1 | XP_016873714.1 | 11, 12 | 1308 | |||
XM_017018226.1 | XP_016873715.1 | 13, 14 | 1869 | |||
XM_017018227.1 | XP_016873716.1 | 8, 9 | 895 | |||
GenBank mRNA | AB012958.1 | - | 13, 14 | 1491 | ||
AK095352.1 | - | 7, 8 | 1131 | |||
AK222772.1 | - | 13, 14 | 1445 | |||
AK291011.1 | - | 13, 14 | 1532 | |||
AK293782.1 | - | 13, 14 | 1295 | |||
AK296871.1 | - | 7, 8 | 674 | |||
AK299978.1 | - | 1 | 32 | |||
AK316133.1 | - | 7, 8 | 741 | |||
BC064837.1 | - | 13, 14 | 1464 | 6186851 |