Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000225.1 | NP_001000225.1 | 1 | 520 |
Gene Symbol: LOC100912217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748966.3 | XP_003749014.2 | 1 | 533 |
Gene Symbol: LOC100912408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748968.3 | XP_003749016.2 | 1 | 533 |
Gene Symbol: Olr282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000224.1 | NP_001000224.1 | 1 | 556 |