Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303543.1 | NP_001290472.1 | 5 | 636 | ||
NM_001303544.1 | NP_001290473.1 | 6 | 574 | |||
NM_001303545.1 | NP_001290474.1 | 5 | 481 | |||
NM_003372.6 | NP_003363.1 | 5 | 485 | |||
GenBank mRNA | AK301646.1 | - | 6 | 574 | ||
AK313420.1 | - | 5 | 453 | |||
BC046094.1 | - | 5 | 438 | 6058145 | ||
BT019604.1 | - | 5 | 426 | |||
CR541932.1 | - | 4 | 315 | |||
DA610631.1 | - | 5 | 485 | |||
U56833.1 | - | 4 | 334 | |||
U96759.1 | - | 5 | 633 | |||
Y17394.1 | - | 5 | 438 |