Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006971.1 | NP_001006972.1 | 12 | 1045 | ||
XM_006230228.2 | XP_006230290.1 | 13 | 1742 | |||
XM_008759871.1 | XP_008758093.1 | 13 | 1853 | |||
XM_008759872.1 | XP_008758094.1 | 14 | 1347 | |||
XM_008759873.1 | XP_008758095.1 | 14 | 1422 | |||
XM_008759874.1 | XP_008758096.1 | 14 | 1659 | |||
XM_008759875.1 | XP_008758097.1 | 14 | 1799 | |||
GenBank mRNA | BC083704.1 | - | 12 | 1045 | 7190222 |