Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135698.1 | NP_001129170.1 | 2 | 208 | ||
XM_006230233.1 | XP_006230295.1 | 2 | 485 | |||
XM_006230235.1 | XP_006230297.1 | 2 | 174 | |||
XM_008759876.1 | XP_008758098.1 | 2 | 485 | |||
GenBank mRNA | CB582520.1 | - | 2 | 220 | ||
FQ229460.1 | - | 2 | 197 | |||
FQ229697.1 | - | 2 | 197 |