Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078.3 | NP_001069.1 | 2 | 518 | ||
NM_080682.2 | NP_542413.1 | 2 | 518 | |||
GenBank mRNA | AK223266.1 | - | 2 | 417 | ||
AK291732.1 | - | 2 | 416 | |||
AU121762.1 | - | 2 | 418 | |||
BC017276.2 | - | 2 | 398 | 4845424 | ||
BC068490.1 | - | 2 | 416 | 30339170 | ||
BC085003.1 | - | 2 | 398 | 6375943 | ||
DB096825.1 | - | 2 | 518 | |||
M30257.1 | - | 2 | 403 | |||
M60335.1 | - | 2 | 396 | |||
X53051.1 | - | 2 | 297 |