Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000243.1 | NP_001000243.1 | 1 | 458 |
Gene Symbol: Olr306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000766.1 | NP_001000766.1 | 1 | 458 |