Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005429.4 | NP_005420.1 | 6 | 1517 | ||
GenBank mRNA | AK313879.1 | - | 6 | 1283 | ||
BC035212.1 | - | 6 | 1348 | 4753457 | ||
BC063685.1 | - | 6 | 1499 | 4513934 | ||
U43142.1 | - | 6 | 1454 | |||
U58111.1 | - | 6 | 1406 | |||
X94216.1 | - | 6 | 1434 |