Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111077.1 | NP_001104547.1 | 6, 7 | 727 | ||
NM_003379.4 | NP_003370.2 | 5, 6 | 761 | |||
XM_011536110.1 | XP_011534412.1 | 2, 3 | 138 | |||
GenBank mRNA | AK296738.1 | - | 6, 7 | 556 | ||
AK299456.1 | - | 4, 5 | 590 | |||
AK312597.1 | - | 6, 7 | 671 | |||
AL162086.1 | - | 6, 7 | 679 | |||
BC013903.2 | - | 5, 6 | 678 | 2959399 | ||
BC068458.1 | - | 5, 6 | 687 | 4819793 | ||
FJ224318.1 | - | 5, 6 | 663 | |||
J05021.1 | - | 5, 6 | 622 | |||
X51521.1 | - | 5, 6 | 663 |